2-Chloro-1,4-benzoquinone, 95%
2-Chloro-1,4-benzoquinone, 95%
2-Chloro-1,4-benzoquinone, 95%
2-Chloro-1,4-benzoquinone, 95%
Thermo Scientific Chemicals

2-Chloro-1,4-benzoquinone, 95%

CAS: 695-99-8 | C6H3ClO2 | 142.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 331220250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS695-99-8
IUPAC Name2-chlorocyclohexa-2,5-diene-1,4-dione
Molecular FormulaC6H3ClO2
InChI KeyWOGWYSWDBYCVDY-UHFFFAOYSA-N
SMILESClC1=CC(=O)C=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=94 %
Melting point52°C to 57°C
Appearance (Color)Yellow to brown
Appearance (Form)Crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only