2'-Hydroxy-5'-methoxyacetophenone, 99%
2'-Hydroxy-5'-methoxyacetophenone, 99%
2'-Hydroxy-5'-methoxyacetophenone, 99%
2'-Hydroxy-5'-methoxyacetophenone, 99%
Thermo Scientific Chemicals

2'-Hydroxy-5'-methoxyacetophenone, 99%

CAS: 705-15-7 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 331240250
Price (EUR)
162,00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
162,00
Each
Add to cart
Chemical Identifiers
CAS705-15-7
IUPAC Name1-(2-hydroxy-5-methoxyphenyl)ethan-1-one
Molecular FormulaC9H10O3
InChI KeyMLIBGOFSXXWRIY-UHFFFAOYSA-N
SMILESCOC1=CC=C(O)C(=C1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point48°C to 51°C
GC>=98.5 %
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only