5-Bromo-2-hydroxybenzyl alcohol, 98%
5-Bromo-2-hydroxybenzyl alcohol, 98%
5-Bromo-2-hydroxybenzyl alcohol, 98%
Thermo Scientific Chemicals

5-Bromo-2-hydroxybenzyl alcohol, 98%

CAS: 2316-64-5 | C7H7BrO2 | 203.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 331480250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS2316-64-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or flakes
GC>=97.5 %
Appearance (Color)White to light brown
Infrared spectrumConforms
Melting point106°C to 111°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only