(S)-(-)-2-Methoxypropionic acid, 98%
(S)-(-)-2-Methoxypropionic acid, 98%
(S)-(-)-2-Methoxypropionic acid, 98%
Thermo Scientific Chemicals

(S)-(-)-2-Methoxypropionic acid, 98%

CAS: 23953-00-6 | C4H8O3 | 104.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
331725000500 mgGlass bottle
Catalog number 331725000
Price (USD)
-
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS23953-00-6
IUPAC Name(2S)-2-methoxypropanoate
Molecular FormulaC4H7O3
InChI KeyICPWFHKNYYRBSZ-VKHMYHEASA-M
SMILESCO[C@@H](C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Specific optical rotation-70° to -80° (20°C, 589 nm) (neat)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only