Nunc™ Universal Latch Rack for Cryogenic Tubes
Nunc™ Universal Latch Rack for Cryogenic Tubes
Nunc™ Universal Latch Rack for Cryogenic Tubes
Nunc™ Universal Latch Rack for Cryogenic Tubes
Thermo Scientific™

Nunc™ Universal Latch Rack for Cryogenic Tubes

Store tubes for automated picking/placing, liquid handling and other high throughput applications using Nunc Universal Latch Rack for Cryogenic Tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHolds
3318252 mL Cryogenic Tubes
3318265 mL Cryogenic Tubes
3318271 to 2 mL Cryogenic Tubes
3318283.6 to 5 mL Cryogenic Tubes
Catalog number 331825
Price (USD)
183.61
Online Exclusive
214.17
Save 30.56 (14%)
Case of 10
Holds:
2 mL Cryogenic Tubes
Price (USD)
183.61
Online Exclusive
214.17
Save 30.56 (14%)
Case of 10

Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes. Features a transparent cover with options for tubes of several sizes.

Ordering Information:

  • Non-sterile
  • Case of 10
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
For Use With (Equipment)For 2mL Cryobank, non-sterile
Holds2 mL Cryogenic Tubes
Quantity10/pk., 10/cs.
ColorTransparent
TypeCryogenic Tube Storage Rack (w/Latch)
Unit SizeCase of 10