Nunc™ Universal Latch Rack for Cryogenic Tubes
Nunc™ Universal Latch Rack for Cryogenic Tubes
Nunc™ Universal Latch Rack for Cryogenic Tubes
Nunc™ Universal Latch Rack for Cryogenic Tubes
Thermo Scientific™

Nunc™ Universal Latch Rack for Cryogenic Tubes

Store tubes for automated picking/placing, liquid handling and other high throughput applications using Nunc Universal Latch Rack for Cryogenic Tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHolds
3318271 to 2 mL Cryogenic Tubes
3318252 mL Cryogenic Tubes
3318265 mL Cryogenic Tubes
3318283.6 to 5 mL Cryogenic Tubes
Catalog number 331827
Price (USD)
182.70
Online Exclusive
213.15
Save 30.45 (14%)
Each
Holds:
1 to 2 mL Cryogenic Tubes
Price (USD)
182.70
Online Exclusive
213.15
Save 30.45 (14%)
Each

Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific™ Nunc™ Universal Latch Rack for Cryogenic Tubes. Features a transparent cover with options for tubes of several sizes.

Ordering Information:

  • Non-sterile
  • Case of 10
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Specifications
For Use With (Equipment)1-2mL Standard Cryotubes, non-sterile
Holds1 to 2 mL Cryogenic Tubes
Quantity10/pk., 10/cs.
ColorTransparent
TypeCryogenic Tube Storage Rack (w/Latch)
Unit SizeEach