(S)-(+)-2-Methoxypropanol, 97%
(S)-(+)-2-Methoxypropanol, 97%
(S)-(+)-2-Methoxypropanol, 97%
Thermo Scientific Chemicals

(S)-(+)-2-Methoxypropanol, 97%

CAS: 116422-39-0 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
2.5 mL
10 mL
Packaging:
Glass bottle
Catalog number 331850010
Price (USD)
-
Quantity:
1 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS116422-39-0
IUPAC Name(2S)-2-methoxypropan-1-ol
Molecular FormulaC4H10O2
InChI KeyYTTFFPATQICAQN-BYPYZUCNSA-N
SMILESCO[C@@H](C)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Specific optical rotation+38° to +44° (20°C, 589 nm) (c=1.66, CHCl3)
Water=<0.1 % (K.F.)
Infrared spectrumConforms
Refractive index1.4060 to 1.4090 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only