CultiMaxx™ Shelving System for G-Rex 500M-CS Bioreactors
Thermo Scientific™
CultiMaxx™ Shelving System for G-Rex 500M-CS Bioreactors
CultiMaxx™ Shelving Systems to support G-Rex 500M-CS Bioreactors
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of ShelvesMaterial
332342G013Stainless Steel
332395G087Stainless Steel
501647813Stainless Steel (18/8)
Catalog number 332342G01
Price (USD)
-
No. of Shelves:
3
Material:
Stainless Steel
Maximize the capacity, increase throughput and optimize cell growth in your lab using the CultiMaxx shelving system in a range of large capacity CO2 Incubators.
Maximize incubator space  
  • These shelving systems are designed to support scale-out of the G-Rex 500M-CS* in cell therapy production.
  • With an in-depth review of the shelving design, this new system provides up to a 150% increase in capacity for the G-Rex, or another way to look at it: 2 1/2 incubators in 1.
 Easy installation
  • Shelving systems are retrofittable and can be self-installed.
*G-Rex is a registered trademark of Wilson Wolf Corporation. G-Rex bioreactors must be ordered from Wilson Wolf Corporation.
Specifications
DescriptionShelving System to support G-Rex™ for Forma™ Steri-Cult™ 3310/3311 323L CO2 Incubator
For Use With (Equipment)Forma Steri-Cult 3310/3311 CO2 Incubator
Includes3 shelves
TypeShelf
MaterialStainless Steel
No. of Shelves3
Unit SizeEach