CultiMaxx™ Shelving Systems for Cell Culture Bags
CultiMaxx™ Shelving Systems for Cell Culture Bags
CultiMaxx™ Shelving Systems for Cell Culture Bags
Thermo Scientific™
CultiMaxx™ Shelving Systems for Cell Culture Bags
CultiMaxx™ Shelving System to support Cell Culture Bags
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoFor Use With (Equipment)No. of Shelves
332474G01Forma Steri-Cult CO2 Incubator8
50166275Heracell VIOS 250i CO2 Incubator8
332475G01Reach-In 821L CO2 Incubator15
Número do catálogo 332474G01
Preço (BRL)
-
For Use With (Equipment):
Forma Steri-Cult CO2 Incubator
No. of Shelves:
8
Maximize the capacity, increase throughput and optimize cell growth in your lab using the CultiMaxx™ Shelving Systems in a range of large capacity CO2 Incubators.
These shelving systems are designed to support scale-out of cell culture bags in cell therapy production. With an in-depth review of the shelving design, this new system provides up to 3X more vessel capacity per unit. The shelving is specifically designed to promote greater gas exchange with increased exposure to the bottom of the semi-permeable bag. Since they are gas permeable on both sides, CultiMaxx shelving for cell culture bags will increase gas exchange compared to standard shelving. These shelves extend to support individual access and improved ease of handling.
Especificações
DescriptionShelving system to support Cell Culture Bags with base set for Forma™ Steri-Cult™ 3310/3311 323L CO2 Incubator
For Use With (Equipment)Forma Steri-Cult CO2 Incubator
Includes8 shelves
TypeShelving System
MaterialStainless Steel
No. of Shelves8
Unit SizeEach