Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)
Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)
Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)
Thermo Scientific Chemicals

Carboxymethyl cellulose, sodium salt, average M.W. 250000 (DS=0.7)

CAS: 9004-32-4 | (C12 H14 O9 R6)n | 263.20 g/mol
数量:
100 g
1 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 332610010
価格(JPY)
-
数量:
1 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9004-32-4
IUPAC Name2,3,4,5,6-pentahydroxyhexanal acetic acid sodium
Molecular Formula(C12 H14 O9 R6)n
InChI KeyDPXJVFZANSGRMM-UHFFFAOYNA-N
SMILES[Na].CC(O)=O.OCC(O)C(O)C(O)C(O)C=O
さらに表示
Appearance (Color)White to beige
Infrared spectrumConforms
Loss on drying=<10 % (As packed) (3 to 5 g, 105°C, 2 h)
Mercury (Hg)=<1 ppm
Degree of substitution0.65 to 0.90
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only