Sodium caprylate, 98%
Sodium caprylate, 98%
Sodium caprylate, 98%
Sodium caprylate, 98%
Thermo Scientific Chemicals

Sodium caprylate, 98%

CAS: 1984-06-1 | C8H15NaO2 | 166.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 334605000
Price (USD)
273.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
273.00
Each
Chemical Identifiers
CAS1984-06-1
IUPAC Namesodium octanoate
Molecular FormulaC8H15NaO2
InChI KeyBYKRNSHANADUFY-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=97.5 % (on dry substance)
Appearance (Form)Powder
Water=<5 % (K.F.)
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only