2-Amino-4,5-dimethoxybenzoic acid, 98%
2-Amino-4,5-dimethoxybenzoic acid, 98%
2-Amino-4,5-dimethoxybenzoic acid, 98%
Acros Organics

2-Amino-4,5-dimethoxybenzoic acid, 98%

CAS: 5653-40-7 | C9H11NO4 | 197.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 335010050
also known as 33501
HSN 29225090
Price (INR)
5,366.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,366.00
Each
Chemical Identifiers
CAS5653-40-7
IUPAC Name2-amino-4,5-dimethoxybenzoic acid
Molecular FormulaC9H11NO4
InChI KeyHJVAVGOPTDJYOJ-UHFFFAOYSA-N
SMILESCOC1=CC(N)=C(C=C1OC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only