Ammonium iron(III) oxalate trihydrate, 98%, pure
Ammonium iron(III) oxalate trihydrate, 98%, pure
Ammonium iron(III) oxalate trihydrate, 98%, pure
Ammonium iron(III) oxalate trihydrate, 98%, pure
Thermo Scientific Chemicals

Ammonium iron(III) oxalate trihydrate, 98%, pure

CAS: 13268-42-3 | C6H18FeN3O15 | 428.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 335100051
Price (EUR)
191,00
Each
Quantity:
5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (EUR)
191,00
Each
Chemical Identifiers
CAS13268-42-3
IUPAC Nametriammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4⁵.4⁵]tridecane-5,5,5-tris(ylium) trihydrate
Molecular FormulaC6H18FeN3O15
InChI KeyARJSTCBPHIPXIJ-UHFFFAOYSA-K
SMILES[NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with KMnO497.5 to 102.5 %
Appearance (Color)Green
Appearance (Form)or fused solid
Appearance (Form)Adhering crystals or crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only