3-Methyl-1-pentanol, 99+%
3-Methyl-1-pentanol, 99+%
3-Methyl-1-pentanol, 99+%
Thermo Scientific Chemicals

3-Methyl-1-pentanol, 99+%

CAS: 589-35-5 | C6H14O | 102.18 g/mol
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 335320250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS589-35-5
IUPAC Name3-methylpentan-1-ol
Molecular FormulaC6H14O
InChI KeyIWTBVKIGCDZRPL-UHFFFAOYNA-N
SMILESCCC(C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4190 to 1.4210 (20°C, 589 nm)
GC>=99.0 %
Infrared spectrumConforms
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only