D-Luciferin sodium salt monohydrate, 98+%
D-Luciferin sodium salt monohydrate, 98+%
D-Luciferin sodium salt monohydrate, 98+%
Thermo Scientific Chemicals

D-Luciferin sodium salt monohydrate, 98+%

CAS: 103404-75-7 | C11H7N2NaO3S2 | 302.298 g/mol
Have Questions?
Catalog NumberQuantityPackaging
33678050050 mgGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 336780500
Price (USD)
802.00
Each
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
802.00
Each
Chemical Identifiers
CAS103404-75-7
IUPAC Namesodium 2-(6-oxo-2,6-dihydro-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylate
Molecular FormulaC11H7N2NaO3S2
InChI KeyBZNVUYVALNTPBG-UHFFFAOYNA-M
SMILES[Na+].[O-]C(=O)C1CSC(N1)=C1SC2=CC(=O)C=CC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.0 %
Appearance (Color)Yellow
Infrared spectrumConforms
Appearance (Form)Powder
Water=<7 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only