4-Amino-6-methoxypyrimidine, 97%
4-Amino-6-methoxypyrimidine, 97%
4-Amino-6-methoxypyrimidine, 97%
Thermo Scientific Chemicals

4-Amino-6-methoxypyrimidine, 97%

CAS: 696-45-7 | C5H7N3O | 125.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Catalog number 337450250
Price (TWD)
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS696-45-7
IUPAC Name6-methoxypyrimidin-4-amine
Molecular FormulaC5H7N3O
InChI KeyVELRBZDRGTVGGT-UHFFFAOYSA-N
SMILESCOC1=CC(N)=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder
Titration with HClO4>=96.0 %
Melting point154°C to 159°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only