Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Thermo Scientific™
Nunc™ Biobanking and Cell Culture Cryogenic Tubes
Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCapacidad (métrico)Color del tapónForma del fondo
337516ORG4.5 mLOrangeRound (Starfoot)
337516RED4.5 mLRedRound (Starfoot)
337516YEL4.5 mLYellowRound (Starfoot)
337516GRN4.5 mLGreenRound (Starfoot)
337516BLU4.5 mLBlueRound (Starfoot)
337516PUR4.5 mLPurpleRound (Starfoot)
375353RED1 mLRedConical (Starfoot)
375353ORG1 mLOrangeConical (Starfoot)
375353YEL1 mLYellowConical (Starfoot)
375353GRN1 mLGreenConical (Starfoot)
375353BLU1 mLBlueConical (Starfoot)
375353PUR1 mLPurpleConical (Starfoot)
375418RED1.8 mLRedRound (Starfoot)
375418ORG1.8 mLOrangeRound (Starfoot)
375418YEL1.8 mLYellowRound (Starfoot)
375418GRN1.8 mLGreenRound (Starfoot)
375418BLU1.8 mLBlueRound (Starfoot)
375418PUR1.8 mLPurpleRound (Starfoot)
Número de catálogo 337516ORG
Precio (MXN)
-
Capacidad (métrico):
4.5 mL
Color del tapón:
Orange
Forma del fondo:
Round (Starfoot)
Thermo Scientific™ Nunc Cryotubes and accessories are designed to work together and provide the best solutions for the most trusted sample storage applications.
  • Non-cytotoxic, non-mutatgenic raw materials and pyrogen-free IVD compatible tubes for added sample security
  • External or internally threaded options available
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • Bulk, uncapped, non-sterile versions available for less demanding applications
Especificaciones
Color del tapónOrange
GraduacionesYes
ModeloWriting Surface, A-R Base
Esterilizables en autoclaveNot Autoclavable
Forma del fondoRound (Starfoot)
Capacidad (métrico)4.5 mL
GraduatedYes
MaterialPolypropylene
Tipo de productoCryogenic Storage Tube
EsterilidadSterile
Estilo de roscaExternal
Unit SizeCase of 1200