Dichlorodiethylsilane, 97%
Dichlorodiethylsilane, 97%
Dichlorodiethylsilane, 97%
Thermo Scientific Chemicals

Dichlorodiethylsilane, 97%

CAS: 1719-53-5 | C4H10Cl2Si | 157.11 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 338100250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1719-53-5
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light brown
Infrared spectrumConforms
Refractive index1.4290 to 1.4320 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only