Dimethyl 2-hydroxyethylphosphonate, 95%, Thermo Scientific Chemicals
Dimethyl 2-hydroxyethylphosphonate, 95%, Thermo Scientific Chemicals
Dimethyl 2-hydroxyethylphosphonate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl 2-hydroxyethylphosphonate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 338160250
Price (EUR)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54731-72-5
IUPAC Namedimethyl (2-hydroxyethyl)phosphonate
Molecular FormulaC4H11O4P
InChI KeyTZPPDWDHNIMTDQ-UHFFFAOYSA-N
SMILESCOP(=O)(CCO)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4410 to 1.4460 (20°C, 589 nm)
GC>=94 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only