Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride, 60% in methanol
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride, 60% in methanol
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride, 60% in methanol
Thermo Scientific Chemicals

Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride, 60% in methanol

CAS: 27668-52-6 | C26H58ClNO3Si | 496.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
338531000100 mLGlass bottle
Catalog number 338531000
Price (EUR)
104,00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
104,00
Each
Chemical Identifiers
CAS27668-52-6
IUPAC Namedimethyl(octadecyl)[3-(trimethoxysilyl)propyl]azanium chloride
Molecular FormulaC26H58ClNO3Si
InChI KeyWSFMFXQNYPNYGG-UHFFFAOYSA-M
SMILES[Cl-].CCCCCCCCCCCCCCCCCC[N+](C)(C)CCC[Si](OC)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Titration Argentometric>=58 %
Appearance (Color)Clear yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only