Search
View additional product information for OPD Substrate Tablets (o-phenylenediamine dihydrochloride) - FAQs (34006)
1 product FAQs found
OPD Substrate Tablets are covered under our general 1-year warranty and are guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf).