Nalgene™ Polycarbonate InVitro™ Biotainer™ Bottles and Carboys, 5 L, square - FAQs
View additional product information for Nalgene™ Polycarbonate Biotainer™ Bottles and Carboys - FAQs (3500-20, 3405-42, 3410-08, 3500-05, 3410-42, 3030-42, 3233-42, 3405-16, 3423-42, 3120-42)
1 product FAQs found
What is the shelf life of Nalgene Polycarbonate Biotainer Bottles and Carboys?
The Nalgene Polycarbonate Biotainer Bottles and Carboys have a shelf life of 5 years from the date of manufacture, as long as they are stored correctly and the integrity of the packaging is not compromised. We recommend storing the bottles and carboys in the original packaging at ambient temperature, away from direct sunlight and high humidity.