Pierce™ Stable Peroxide Substrate Buffer (10X)
Thermo Scientific™
Pierce™ Stable Peroxide Substrate Buffer (10X)
Thermo Scientific Pierce Stable Peroxide Substrate Buffer (10X) include optimized, reliable and convenient stock solutions of peroxide for HRP-based assaysRead more
Catalog NumberQuantity
34062100 mL
Catalog number 34062
Price (GBP)
88.75
Each
Quantity:
100 mL
Request bulk or custom format
Price (GBP)
88.75
Each
Thermo Scientific Pierce Stable Peroxide Substrate Buffer (10X) include optimized, reliable and convenient stock solutions of peroxide for HRP-based assays detection.
Features of Stable Peroxide Substrate Buffer (10X):
• Ready-to-use
• Minimize assay-to-assay variability
Buffered stable hydrogen peroxide (H2O2) is used by researchers who prefer to make their own HRP-ELISA substrates. This is easily done by adding a chromogen of choice to the buffer. Stable Peroxide Substrate Buffer has a long shelf life (12 months after receipt) and is provided as a 10-fold concentrate. A total volume of 1000 mL can be made from one 100 mL bottle of concentrate. If you use 10 mL of substrate per plate, this will allow you to make substrate for 100 plates from only one bottle of Stable Peroxide Substrate Buffer.
Related Products
Pierce™ Diethanolamine Substrate Buffer (5X)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionPierce Stable Peroxide Buffer, 10X
For Use With (Application)ELISA, Immunohistochemistry (IHC), Western Blot
FormLiquid
Product LinePierce
Substrate TypeAlkaline Phosphatase, Horseradish Peroxidase
Product TypeELISA Substrate
Quantity100 mL
Unit SizeEach
Contents & Storage
Upon receipt store product at 4°C. Product is shipped at ambient temperature.