Metal Enhanced DAB Substrate Kit

Catalog number: 34065B

Thermo Scientific™  Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce Metal-Enhanced DAB Substrate Kit uses cobalt and nickel chloride in a special formulation of diaminobenzidine peroxidase substrate that yields intense color for immunohistochemical staining.

Features of the Metal-Enhanced DAB Substrate Kit :

Incredible sensitivity—fifty times more sensitive than the traditional DAB method and thiry times more sensitive than other metal-intensified versions of DAB
Low background, high intensity—get a crisp dark brown-black precipitate that is more intense than the dull brown precipitate when using DAB without enhancement. And, even with the increased intensity, background is almost nonexistent
Only two components—simply mix the two liquid components and your working solution is ready to use
Six-hour stability—the innovative working solution is stable for more than six hours at room temperature, while other DAB substrates must be used immediately

To prepare the product, the 10X Metal Enhanced Substrate Solution is diluted with the Pierce Stable Peroxide Buffer prior to use, and the special formulation allows the 1X substrate to be stable for at least 6 hours at room temperature. The background color of the substrate does not increase and, unlike typical metal enhanced DAB methods, the reactivity is unchanged (1,2). The resulting precipate is a dark brown-black that is more intense than the brown color from DAB alone. The intensity not only increases, but the background is also reduced, which increases the signal-to-noise ratio and makes screening tissue or membrane antigens easier.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Chromogenic, Colorimetric
Product Line: Pierce™
Product Size: Custom
Substrate: DAB (Enhanced)
Substrate Type: HRP (Horseradish Peroxidase) Substrate
Technique: IHC (Immunohistochemistry), Immunodetection

Contents & storage

• Metal Enhanced DAB (10X) (store at -20°C)
• Stable Peroxide Buffer (store at 4°C)

Documents

Manuals & protocols