SuperSignal™ West Dura Extended Duration Substrate
Thermo Scientific™

SuperSignal™ West Dura Extended Duration Substrate

Luminol-based enhanced chemiluminescence (ECL) horseradish peroxidase (HRP) substrate ideal for quantitative western blotting with stable light output for mid-femtogram level detection for western blot analysis.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
34075X44 x 100 mL
34075100 mL
34076200 mL
34076X44 x 200 mL
3707120 mL
카탈로그 번호 34075X4
제품 가격(KRW)
2,242,000
Each
수량:
4 x 100 mL
제품 가격(KRW)
2,242,000
Each

Thermo Scientific SuperSignal West Dura Extended Duration Substrate is a luminol-based enhanced chemiluminescence (ECL) horseradish peroxidase (HRP) substrate ideal for quantitative western blotting with stable light output for mid-femtogram level detection for western blot analysis.

SuperSignal West Dura Extended Duration Substrate characteristics include:

  • Sensitivity: mid-femtogram
  • Stability: 24-hr working solution stability; 1-year kit stability at room temperature
  • Compatibility: nitrocellulose and PVDF membranes
  • Signal duration: 24 hours
  • Recommended primary antibody concentration: 1:1,000–1:50,000 dilution (0.02–1.0 μg/mL)
  • Recommended secondary antibody concentration: 1:50,000–1:250,000 dilution (4–20 ng/mL)

SuperSignal West Dura Extended Duration Substrate for HRP is optimized for high sensitivity and long signal duration, making it ideal for cooled CCD camera-based detection systems. Unlike substrates with signals that decline to barely detectable levels in 30–60 minutes, the signal produced with SuperSignal West Dura chemiluminescent substrate is stable for 24 hours, allowing multiple film or camera exposures.

For Research Use Only. Not for use in diagnostic procedures.

General References:

  1. Gangalum, R.K.et al.(2004).J. Biol. Chem. 279,43374-43377.
  2. MacInnis, B.L. and Campenot, R.B. (2004).Science 295,1536-1539.
  3. Sato, S.,et al.(2003).J. Biol. Chem. 278,15123-15127.
  4. Tokumaru, H.,et al.(2001).Cell 104,421-432.
사양
권장 항체 농도2°: 1:50-250 K (4-20 ng/mL), 1°: 1:5 K (0.02-1 μg/mL)
민감도Mid-femtogram
신호 지속 시간24 hr.
검출 방법Chemiluminescence
형태Liquid
제품라인SuperSignal
제품 유형Substrate
수량4 x 100 mL
SubstrateHorseradish Peroxidase
Unit SizeEach
구성 및 보관
Sufficient For: 48 mini-gel blots; 4000 cm2 of membrane
• Luminol/Enhancer, 4 x 50 mL
• Stable Peroxide Buffer, 4 x 50 mL

Store at room temperature.