CL-XPosure™ Film, 5 x 7 in. (12.5 x 17.5 cm)

Catalog number: 34088

Thermo Scientific™  Related applications: Protein Assays and Analysis | Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific CL-XPosure Film is an economically priced clear-blue X-ray film especially for detection and documentation of ECL-based Western blots and other chemiluminescent protein or nucleic acid assay methods.

CL-XPosure Film is an excellent phototgraphic film for use with enhanced chemiluminescence (ECL) substrates for horseradish peroxidase (HRP) or alkaline phosphatase (AP). Convenient sizes are offered for protein Western blotting, standard film cassettes and film developing equipment. The autoradiography film also works well for chemiluminescent or isotopic DNA and RNA methods, including Southern and Northern blotting and gel-shift assays (EMSA). The quality and price of this film make it the smart choice for life science research applications.

Features of CL-XPosure Film:

High performance – just as sensitive as other commercially available autoradiography films
Affordable – costs up to 75% less than other brands of comparable quality X-ray film
Convenient – available in five sheet sizes in packages containing 50 or 100 non-interleaved sheets
Perfect match – validated for use with Thermo Scientific SuperSignal Chemiluminescent Substrates and Thermo Scientific Pierce ECL Chemiluminescent Substrate

Related Products
CL-XPosure™ Film, 5 x 7 in. (13 x 18 cm)
CL-XPosure™ Film, 7 x 9.5 in. (18 x 24 cm)
CL-XPosure™ Film, 8 x 10 in. (20 x 25 cm)
CL-XPosure™ Film, 9.5 x 11.8 in. (24 x 30 cm)
CL-XPosure™ Film, 14 x 17 in. (35 x 43 cm)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Autoradiography
Product Line: CL-Xposure™
Product Size: 50 sheets
Type: Film

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products