Attention: Product will be discontinued in June 2027 or while supplies last.
CL-XPosure™ Film
Thermo Scientific™

CL-XPosure™ Film

Thermo Scientific CL-XPosure Film is an economically priced clear-blue X-ray film especially for detection and documentation of ECL-based western blots and other chemiluminescent protein or nucleic acid assay methods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (LxW)
3409018 x 13 cm
3408924 x 18 cm
Catalog number 34090
Price (USD)
225.65
Online exclusive
238.00
Save 12.35
100 sheets
Dimensions (LxW):
18 x 13 cm
Price (USD)
225.65
Online exclusive
238.00
Save 12.35
100 sheets
Thermo Scientific CL-XPosure Film is an economically priced clear-blue X-ray film especially for detection and documentation of ECL-based western blots and other chemiluminescent protein or nucleic acid assay methods. CL-XPosure Film is an excellent photographic film for use with enhanced chemiluminescence (ECL) substrates for horseradish peroxidase (HRP) or alkaline phosphatase (AP). Convenient sizes are offered for protein western blotting, standard film cassettes, and film developing equipment. This autoradiography film also works well for chemiluminescent or isotopic DNA and RNA methods, including Southern and northern blotting and gel-shift assays (EMSA). The quality and price of this film make it the ideal choice for life science research applications.

Features include:
• High performance—just as sensitive as other commercially available autoradiography films
• Economical—achieve high-quality images at an affordable price
• Convenient—available in five sheet sizes in packages containing 100 non-interleaved sheets
• Perfect match—validated for use with SuperSignal chemiluminescent substrates and Pierce ECL and ECL Plus chemiluminescent substrates

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity100 Sheets
Dimensions (LxW)18 x 13 cm
Length (English)7 in.
Product LineCL-Xposure
Product TypeX-Ray Film
Width (English)5 in.
Unit Size100 sheets
Contents & Storage
Store at room temperature.