Temperature Flow and Pressure Control Accessories
Thermo Scientific™
Temperature Flow and Pressure Control Accessories
Refine the pressure and flow characteristics of your Thermo Scientific™ recirculating chiller.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)Size
342000000025Pressure ReducerThermoFlex 900 - 100001/2 in. NPT
342000000026Pressure ReducerThermoFlex 3500 - 50003/4 in. NPT
342000000027Pressure ReducerThermoFlex 7500 - 240001 in. NPT
611000000106Flow Control KitThermoFlex 900 - 100001/2 in. NPT
342000000028Flow Control KitThermoFlex 3500 - 50003/4 in. NPT
342000000029Flow Control KitThermoFlex 7500 - 240001 in. NPT
Catalog number 342000000025
Price (CLP)
-
Type:
Pressure Reducer
For Use With (Equipment):
ThermoFlex 900 - 10000
Size:
1/2 in. NPT
Customize the flow and pressure output of recirculating chiller to match the needs of your process. Available in several popular sizes Thermo Scientific™ Flow and pressure control kits for Thermo Scientific chillers can have you up and running without the hassle of finding parts.
  • Designed to work with your Thermo Scientific recirculating chiller
  • Available in popular sizes that match your recirculating chiller connections and interface with your process
  • Take the guess work out of finding parts to match your recirculating chiller connections
  • Designed to match the operating range of your recirculating chiller
Additional questions? Please contact Thermo Scientific Technical support if you’re unsure how best to refine the flow and pressure of your recirculating chiller.
Specifications
For Use With (Equipment)ThermoFlex 900 - 10000
Pump TypeCentrifugal Pump, Turbine Pump
Size1/2 in. NPT
TypePressure Reducer
Unit SizeEach