Nalgene™ Square PETG Media Bottles with Closure: Sterile, Shrink-Wrapped Trays, 250mL - FAQs
Nalgene™ Square PETG Media Bottles with Closure: Sterile, Shrink-Wrapped Trays, 250mL - FAQs
View additional product information for Nalgene™ Square PETG Media Bottles with Closure: Sterile, Shrink-Wrapped Trays - FAQs (342020-0500, 342020-0125, 342020-9500, 342020-0060, 342020-0030, 342020-0250, 342020-2000, 342020-1000)
2 product FAQs found
What is the level of accuracy for the gradations on the Nalgene Square PETG Media Bottles with Closure: Sterile, Shrink-Wrapped Trays?
The lines on our Nalgene bottles are not tested to any level of accuracy and are intended for reference only.
What is the difference between the following Nalgene Square PETG Media Bottles: Cat. No. 2019-xxxx (with Closure), Cat. No. 342020-xxxx (with Closure: Sterile, Shrink-Wrapped Trays), Cat. No. CE-N2019-xxxx (Platinum Certified Clean)?
All three products use the same PETG bottles, so the materials and specifications are the same for the bottle itself. Cat. No. 342020-xxxx is supplied with natural-colored HDPE closures, while Cat. No. 2019-xxxx and Cat. No. CE-N2019-xxxx are supplied with white HDPE closures. The HDPE resin used for the closures is the same, but a white colorant has been added to the resin for Cat. No. 2019-xxxx and Cat. No. CE-N2019-xxxx (this will be noted on the COA). Cat. No. 2019-xxxx also has a PVC tamper-resistant seal around the closure, while Cat. No. 342020-xxxx does not.
For more detail about the different "levels of clean" for our Thermo Scientific Nalgene contains, please refer to our Nalgene Clean Containers brochure.