Nalgene™ Continuous-Thread Lined Closures for Serum Vials: Sterile, Bulk Pack
Actual product may vary
Nalgene™ Continuous-Thread Lined Closures for Serum Vials: Sterile, Bulk Pack
Nalgene™ Continuous-Thread Lined Closures for Serum Vials: Sterile, Bulk Pack
Nalgene™ Continuous-Thread Lined Closures for Serum Vials: Sterile, Bulk Pack
Nalgene™ Continuous-Thread Lined Closures for Serum Vials: Sterile, Bulk Pack
Eliminate the need for a rubber stopper with the safe-sealing solution of these continuous thread lined closures for serum vials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure Color
342158-0025Red
342158-0021White
342158-0022Black
342158-0023Yellow
342158-0024Green
342158-0026Blue
Catalog number 342158-0025
HSN 39235090
Price (INR)
143,565.00
Case
Closure Color:
Red
Price (INR)
143,565.00
Case
Eliminate the need for a rubber stopper with the safe-sealing solution of high density polyetylene (HDPE) Thermo Scientific™ Nalgene™ Continuous Thread Lined Closures for Serum Vials.
Features:
  • Sterile to 10-6 SAL
  • Liner made of an Low Density Polyethylene (LDPE) core between two facings of solid HDPE
  • Multiple closure colors available
Ordering Information:
Serum Vial closures are bulk packed at 2,000 per case.
The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database
Specifications
Closure ColorRed
DescriptionSerum vial closures
Material (Liner)Three-ply co-extruded liner (LDPE core between two facing of solid HDPE)
ColorRed
Diameter (Metric) Outer22.1 mm
MaterialHDPE
SterilitySterile
No. per Case2000
No. per Pack2000
Product LineNalgene
Closure MaterialRubber
Closure TypeLined Continuous-thread Closure
Quantity2000 Cs.
Closure Size20-415
For Use With (Equipment)Sterile PETG Serum Vials (Sold Separately)
Certifications/ComplianceISO 13485:2016
Height (Metric)14.2 mm
Product TypeSerum Vial Closure
Unit SizeCase