8-Methylnonanoic acid, 97%
8-Methylnonanoic acid, 97%
8-Methylnonanoic acid, 97%
Thermo Scientific Chemicals

8-Methylnonanoic acid, 97%

CAS: 5963-14-4 | C10H19O2 | 171.26 g/mol
数量:
250 mg
1 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 342342500
価格(JPY)
-
数量:
250 mg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5963-14-4
IUPAC Name8-methylnonanoate
Molecular FormulaC10H19O2
InChI KeyOAOABCKPVCUNKO-UHFFFAOYSA-M
SMILESCC(C)CCCCCCC([O-])=O
さらに表示
GC>=96.0 %
Refractive index1.4330 to 1.4350 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only