Nalgene™ PPCO Micro Packaging Vials with E-beam Irradiation, Sterile, Bulk Pack, 0.5mL capacity, Natural, Sterile - FAQs
View additional product information for Nalgene™ PPCO Micro Packaging Vials with E-beam Irradiation: Sterile, Bulk Pack - FAQs (342810-0020, 342810-0005)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.