Nalgene™ PPCO High-Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack, orange - FAQs
View additional product information for Nalgene™ PPCO High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack - FAQs (342820-0112, 342820-0113, 342820-0110, 342820-0111, 342820-0116, 342820-0117, 342820-0114, 342820-0115, 342820-0118, 342820-0119, 342820-1110)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.