4-Acetylbenzenesulfonyl chloride, 97%
4-Acetylbenzenesulfonyl chloride, 97%
4-Acetylbenzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

4-Acetylbenzenesulfonyl chloride, 97%

CAS: 1788-10-9 | C8H7ClO3S | 218.65 g/mol
Quantity:
5 g
50 g
Packaging:
Glass bottle
Catalog number 343850500
Price (JPY)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1788-10-9
IUPAC Name4-acetylbenzene-1-sulfonyl chloride
Molecular FormulaC8H7ClO3S
InChI KeyFXVDNCRTKXMSEZ-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Additional infoMay contain varying amounts of inorganic chloride
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only