Manganeso(III)acetilacetonato, 97 %, Thermo Scientific Chemicals
Manganeso(III)acetilacetonato, 97 %, Thermo Scientific Chemicals
Manganeso(III)acetilacetonato, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganeso(III)acetilacetonato, 97 %, Thermo Scientific Chemicals

CAS: 14284-89-0 | C15H21MnO6 | 352.27 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
100 g
Envase:
Frasco de vidrio
Número de catálogo 343980050
Precio (EUR)
18,00
Each
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
18,00
Each
Identificadores químicos
CAS14284-89-0
IUPAC Namemanganese(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21MnO6
InChI KeyHYZQBNDRDQEWAN-LNTINUHCSA-K
SMILES[Mn+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Black
Infrared spectrumConforms
Elemental Analysis>=96 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only