Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Thermo Scientific™
Molecular BioProducts Snap-Cap Plain Centrifuge Tubes
Have Questions?
Catalog NumberColorSterilityCapacity (Metric)
3439AAssortedSterile0.6 mL
Catalog number 3439A
Price (USD)
-
Color:
Assorted
Sterility:
Sterile
Capacity (Metric):
0.6 mL
  • With flat cap
  • No frosting or graduations
  • Certified RNase/DNase, RNA/DNA, ATP and pyrogen free
Specifications
Cap StyleFlat
Certifications/ComplianceRNase, DNase, RNA, DNA, ATP, pyrogen-free
IncludesFlat cap
PackagingBag
Capacity (Metric)0.6 mL
Closure TypeFlat Cap
ColorAssorted
GraduatedNo
MaterialPolypropylene
SterilitySterile
Unit SizeEach