Triethylenetetramine, 60%
Triethylenetetramine, 60%
Triethylenetetramine, 60%
Thermo Scientific Chemicals

Triethylenetetramine, 60%

CAS: 112-24-3 | C6H18N4 | 146.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
2.5 kg
Catalog number 344070025
Price (USD)
148.65
Special offer
Online exclusive
Ends: 28-Dec-2026
175.00
Save 26.35
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)
148.65
Special offer
Online exclusive
Ends: 28-Dec-2026
175.00
Save 26.35
Each
Chemical Identifiers
CAS112-24-3
IUPAC Name(2-aminoethyl)({2-[(2-aminoethyl)amino]ethyl})amine
Molecular FormulaC6H18N4
InChI KeyVILCJCGEZXAXTO-UHFFFAOYSA-N
SMILESNCCNCCNCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<150 APHA
Refractive index1.4960 to 1.5000 (20°C, 589 nm)
Infrared spectrumConforms
GC>=55 %
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only