Brij™ C20
Brij™ C20
Brij™ C20
Thermo Scientific Chemicals

Brij™ C20

CAS: 9004-95-9 | C56H114O21 | 1123.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Catalog number 344295000
Price (USD)
89.30
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
89.30
Each
Chemical Identifiers
CAS9004-95-9
MDL NumberMFCD00080892
CAS9004-95-9
MDL NumberMFCD00080892
SpecificationsSpecification SheetSpecification Sheet
Antioxidantwithout Antioxidant
Impurity=<1 ppm free Ethyleneoxide
Antioxidantwithout Antioxidant
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only