6-Amino-2-naphthoic acid, 90%
6-Amino-2-naphthoic acid, 90%
6-Amino-2-naphthoic acid, 90%
Thermo Scientific Chemicals

6-Amino-2-naphthoic acid, 90%

CAS: 116668-47-4 | C11H9NO2 | 187.2 g/mol
Have Questions?
Catalog NumberQuantity
3445000505 g
Catalog number 344500050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS116668-47-4
IUPAC Name6-aminonaphthalene-2-carboxylate
Molecular FormulaC11H8NO2
InChI KeyNZTPZUIIYNYZKT-UHFFFAOYSA-M
SMILESNC1=CC=C2C=C(C=CC2=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Green-beige to brown
Infrared spectrumConforms
HPLC>=88 %
Titration with HClO4>=88 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only