2-Thiopheneboronic acid, 97%
2-Thiopheneboronic acid, 97%
2-Thiopheneboronic acid, 97%
Thermo Scientific Chemicals

2-Thiopheneboronic acid, 97%

CAS: 6165-68-0 | C4H5BO2S | 127.95 g/mol
Quantity:
5 g
25 g
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 344710250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS6165-68-0
IUPAC Name(thiophen-2-yl)boronic acid
Molecular FormulaC4H5BO2S
InChI KeyARYHTUPFQTUBBG-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=96.0 %
Appearance (Color)Off-white to beige
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only