Snap Cap Low Retention Microcentrifuge Tubes
Snap Cap Low Retention Microcentrifuge Tubes
Snap Cap Low Retention Microcentrifuge Tubes
Snap Cap Low Retention Microcentrifuge Tubes
Snap Cap Low Retention Microcentrifuge Tubes
Thermo Scientific™
Snap Cap Low Retention Microcentrifuge Tubes
Improve sample recovery without additives or coatings. These tubes offer maximum sample recovery by significantly reducing surface binding.
Have Questions?
Catalog NumberColor
3448Natural
Catalog number 3448
Price (USD)
218.43
Online Exclusive
286.23
Save 67.80 (24%)
Case of 5000
Color:
Natural
Price (USD)
218.43
Online Exclusive
286.23
Save 67.80 (24%)
Case of 5000
Viscous liquids and costly reagents retained in a microcentrifuge tube effect assay accuracy and precision. Thermo Scientific™ Snap Cap Low Retention Microcentrifuge Tubes improves sample recovery without additives or coatings. The 0.6mL, 1.5mL and 2.0mL snap cap low retention tubes offers maximum sample recovery by significantly reducing surface binding.
  • The duckbill cap reduces the pressure required to open the tube with a wider contact surface yet ensures the cap will remain closed during centrifugation
  • Low retention properties are not achieved by a coating or plastic embedding processes ensuring that samples will not be contaminated from these additives
  • Tubes will remain closed during centrifugation yet open easily with one hand as needed
  • Tubes are available in Amber to protect your light-sensitive samples
  • Graduation marks on the microcentrifuge tube allow for effective sample reference
Color/Packaging Options
  • A variety of packaging options are available to help with contamination control in the lab
  • For stringent sterility requirements, individually wrapped sterile tubes are available
Highest Quality
  • Manufactured from virgin polypropylene these microcentrifuge tubes ensure sample integrity during boiling, freezing, and centrifugation
  • Sterile product is certified free of RNase, DNase, DNA, ATP, bioburden and endotoxins
  • Nonsterile product is certified free of RNase, DNase, DNA and pyrogens
Recommended for:
Daily laboratory usage including centrifugation and short term sample storage.
Specifications
Cap StyleFlat
Certifications/ComplianceCertified free of RNase, DNase, DNA and pyrogens
Description1.5mL Graduated, Nonsterile
Packaging50 tubes/ bag, 10 bags/ unit, 10 units/ case
Capacity (Metric)1.5 mL
ColorNatural
GraduatedYes
MaterialPolypropylene
Max. RCF26,000 x g
SterilityNon-sterile
Unit SizeCase of 5000