Iron(III) sulfate pentahydrate, 97%
Iron(III) sulfate pentahydrate, 97%
Iron(III) sulfate pentahydrate, 97%
Thermo Scientific Chemicals

Iron(III) sulfate pentahydrate, 97%

CAS: 142906-29-4 | Fe2O12S3·5H2O | 489.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 345231000
Price (USD)
52.40
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
52.40
Each
Chemical Identifiers
CAS142906-29-4
IUPAC Namediiron(3+) pentahydrate trisulfate
Molecular FormulaFe2H10O17S3
InChI KeyYHGPYBQVSJBGHH-UHFFFAOYSA-H
SMILESO.O.O.O.O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Subst not pptd by NH3=<0.05 % (as SO4)
Copper (Cu)=<20 ppm
Chloride (Cl)=<50 ppm
Phosphate (PO4)=<30 ppm
Degree of hydrationMay vary
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only