4-(tert-Butyl)benzylbromide, 97%
4-(tert-Butyl)benzylbromide, 97%
4-(tert-Butyl)benzylbromide, 97%
Thermo Scientific Chemicals

4-(tert-Butyl)benzylbromide, 97%

CAS: 18880-00-7 | C11H15Br | 227.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 345500050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS18880-00-7
IUPAC Name1-(bromomethyl)-4-tert-butylbenzene
Molecular FormulaC11H15Br
InChI KeyQZNQSIHCDAGZIA-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration Argentometric>=96.0 %
Infrared spectrumConforms
Refractive index1.5445 to 1.5465 (20°C, 589 nm)
Appearance (Color)Clear light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only