Sodium acetate trihydrate, 99.0-101.0% (dried basis), crystals, USP, Multi-Compendial, J.T.Baker™
Avantor™

Sodium acetate trihydrate, 99.0-101.0% (dried basis), crystals, USP, Multi-Compendial, J.T.Baker™

This product is GMP Manufactured and meets FCC, USP, JP, BP and EP specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
85 lb.
275 lb.
Catalog number 3461-01
Price (BRL)
-
Quantity:
500 g
This product is GMP Manufactured and meets FCC, USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets FCC, USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorColorless
PackagingPoly bottle, tamper-evident cap
Boiling Point120°C to 123°C (1013 hPa)
GradeMulti-Compendial/USP
Molecular Weight (g/mol)136.08
Percent Purity99 to 101%
Physical FormSolid
Quantity500 g
pH7.5 to 9.2 (25°C, 5% solution)
Unit SizeEach