Sodium acetate trihydrate, 99.0-101.0% (dried basis), crystals, USP, Multi-Compendial, J.T.Baker™
Sodium acetate trihydrate, 99.0-101.0% (dried basis), crystals, USP, Multi-Compendial, J.T.Baker™
Avantor™

Sodium acetate trihydrate, 99.0-101.0% (dried basis), crystals, USP, Multi-Compendial, J.T.Baker™

This product is GMP Manufactured and meets FCC, USP, JP, BP and EP specifications.자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
수량:
500 g
2.5 kg
12 kg
85 lb.
275 lb.
카탈로그 번호 3461-05
제품 가격(KRW)
-
견적 요청하기
수량:
2.5 kg
This product is GMP Manufactured and meets FCC, USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
사양
색상Colorless
포장Poly bottle, tamper-evident cap
끓는점120°C to 123°C (1013 hPa)
등급Multi-Compendial/USP
분자량(g/mol)136.08
백분율 순도99 to 101%
물리적 형태Solid
수량2.5 kg
pH7.5 to 9.2 (25°C, 5% solution)
Unit SizeEach