Sodium acetate trihydrate, 99.0-101.0% (dried basis), crystals, USP, Multi-Compendial, J.T.Baker™
Avantor™

Sodium acetate trihydrate, 99.0-101.0% (dried basis), crystals, USP, Multi-Compendial, J.T.Baker™

This product is GMP Manufactured and meets FCC, USP, JP, BP and EP specifications.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
500 g
2.5 kg
12 kg
85 lb.
275 lb.
Número de catálogo 3461-08
Precio (MXN)
-
Cantidad:
85 lb.
This product is GMP Manufactured and meets FCC, USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets FCC, USP, JP, BP and EP specifications.
Especificaciones
CAS6131-90-4
ColorIncolora
EnvasePolyethylene drum, open head
Punto de ebullición120°C to 123°C (1013 hPa)
GradoMulticomponente/USP
Peso molecular (g/mol)136.08
Porcentaje de pureza99 to 101%
Forma físicaSolid
Cantidad85 lb.
pH7.5 to 9.2 (25°C, 5% solution)
Unit SizeEach