Carminic acid, 70-90%
Carminic acid, 70-90%
Carminic acid, 70-90%
Thermo Scientific Chemicals

Carminic acid, 70-90%

CAS: 1260-17-9 | C22H20O13 | 492.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 346630010
Price (EUR)
82,65
Online exclusive
91,60
Save 8,95 (10%)
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
82,65
Online exclusive
91,60
Save 8,95 (10%)
Each
Chemical Identifiers
CAS1260-17-9
IUPAC Name3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[(2R,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
Molecular FormulaC22H20O13
InChI KeyDGQLVPJVXFOQEV-NGOCYOHBSA-N
SMILESCC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C([C@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)=C(O)C(O)=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)Red
HPLC>=70 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only