2,4-hexadiène, 90 %, Tech., mélange d’isomères, Thermo Scientific Chemicals
2,4-hexadiène, 90 %, Tech., mélange d’isomères, Thermo Scientific Chemicals
2,4-hexadiène, 90 %, Tech., mélange d’isomères, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-hexadiène, 90 %, Tech., mélange d’isomères, Thermo Scientific Chemicals

CAS: 592-46-1 | C6H10 | 82.15 g/mol
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
1 g
5 g
Conditionnement:
Flacon en verre
Référence 346670050
Prix (EUR)
132,00
Each
Quantité:
5 g
Conditionnement:
Flacon en verre
Grand volume ou format personnalisé
Prix (EUR)
132,00
Each
Identifiants chimiques
CAS592-46-1
IUPAC Name(2Z,4E)-hexa-2,4-diene
Molecular FormulaC6H10
InChI KeyAPPOKADJQUIAHP-CIIODKQPSA-N
SMILESCC=CC=C/C
Voir plus
Appearance (Form)Liquid
GC>=88 %
Appearance (Color)Clear light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only