Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3
Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3
Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3
Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3
Thermo Scientific Chemicals

Diethyl (3,3-diethoxypropyl)phosphonate, 95%, stabilized with max. 1% w/w Na2CO3

CAS: 15110-17-5 | C11H25O5P | 268.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 346930250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS15110-17-5
IUPAC Namediethyl (3,3-diethoxypropyl)phosphonate
Molecular FormulaC11H25O5P
InChI KeyWKPXMGJOGYEGNW-UHFFFAOYSA-N
SMILESCCOC(CCP(=O)(OCC)OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4290 to 1.4320 (20°C, 589 nm)
GC>=94 %
Appearance (Color)Clear light yellow to orange
Stabilizer=<1 % Na2CO3
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only