Clorhidrato de DL-ornitina, 99 %, Thermo Scientific Chemicals
Clorhidrato de DL-ornitina, 99 %, Thermo Scientific Chemicals
Clorhidrato de DL-ornitina, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Clorhidrato de DL-ornitina, 99 %, Thermo Scientific Chemicals

CAS: 1069-31-4 | C5H13ClN2O2 | 168.62 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
100 g
Número de catálogo 347001000
Precio (USD)
-
Cantidad:
100 g
Pedido a granel o personalizado
Identificadores químicos
CAS1069-31-4
IUPAC Namehydrogen 2,5-diaminopentanoic acid chloride
Molecular FormulaC5H13ClN2O2
InChI KeyGGTYBZJRPHEQDG-UHFFFAOYNA-N
SMILES[H+].[Cl-].NCCCC(N)C(O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
Solubility(10 % in water) Clear colorless to light yellow solution
Appearance (Form)Crystalline powder
Appearance (Color)White to cream
Titration with HClO498.5 to 101.5 %
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only